AC5372911848 Accela ChemBio Inc.

2,3,5,6-Tetrafluoro-4-methylbenzyl Alcohol

  • MDL number MFCD03093260
  • Cas Number : 79538-03-7
  • Molecular Weight : 194.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018258&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018258.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart