AC5372911850 Accela ChemBio Inc.

Ethyl 4-Boc-2-homomorpholinecarboxylate

  • MDL number MFCD21364423
  • Cas Number : 1363166-21-5
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-tert-butyl 2-ethyl 1,4-oxazepane-2,4-dicarboxylate
Inchi Code 1S/C13H23NO5/c1-5-17-11(15)10-9-14(7-6-8-18-10)12(16)19-13(2,3)4/h10H,5-9H2,1-4H3
Inchi Key NFMNWHFTGYMZQU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. -20?
Physical Form Colorless to orange liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018265&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018265.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart