AC5372911851 Accela ChemBio Inc.

Methyl 1-(4-Chlorophenyl)-5-methyl-1,2,3-triazole-4-carboxylate

  • MDL number MFCD12968656
  • Cas Number : 1147199-25-4
  • Molecular Weight : 251.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018266&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018266.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart