AC5372911854 Accela ChemBio Inc.

1-Bromo-2-cyclopropylethyne

  • MDL number MFCD21332903
  • Cas Number : 57951-72-1
  • Molecular Weight : 145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018270&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018270.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart