AC5372911857 Accela ChemBio Inc.

4-Ethylbenzenesulfonamide

  • MDL number MFCD01365828
  • Cas Number : 138-38-5
  • Molecular Weight : 185.24344

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018275&attname=GoodsCode

Safety Information

signalword Warning
pcodes P264-P280-P302+P350-P305+P351+P338-P332+P313-P337+P313-P362
hcodes GHS07,
MSDS http://pdf.accelachem.com/cn/msds/SY018275.pdf
hstatements H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart