AC5372911861 Accela ChemBio Inc.

2,4-Difluoro-3-methylbenzonitrile

  • MDL number MFCD07777155
  • Cas Number : 847502-87-8
  • Molecular Weight : 153.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018279&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280H-P305+P351+P338-P309-P310
hcodes GHS06
MSDS http://pdf.accelachem.com/cn/msds/SY018279.pdf
hstatements H311-H302-H332-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart