AC5372911863 Accela ChemBio Inc.

Ethyl 1-Benzylpiperidine-3-carboxylate

  • MDL number MFCD00191221
  • Cas Number : 72551-53-2
  • Molecular Weight : 247.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018282&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018282.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart