AC5372911888 Accela ChemBio Inc.

N-Boc-3-(4-bromophenyl)cyclobutanamine

  • MDL number MFCD21364424
  • Cas Number : 1363166-45-3
  • Molecular Weight : 326.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018336&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018336.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart