AC5372911889 Accela ChemBio Inc.

4-Boc-4,8-diazabicyclo[5.2.0]nonane

  • MDL number MFCD17015579
  • Cas Number : 1253790-47-4
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018345&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018345.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart