AC5372911900 Accela ChemBio Inc.

3-Amino-N-(2-furylmethyl)-4-methylbenzamide

  • MDL number MFCD09043810
  • Cas Number : 926192-92-9
  • Molecular Weight : 230.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018386&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018386.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart