AC5372911901 Accela ChemBio Inc.

3-Bromo-N-(2-cyanophenyl)-2-methylbenzamide

  • MDL number MFCD20823212
  • Cas Number : 1363165-97-2
  • Molecular Weight : 315.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018387&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018387.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart