AC5372911902 Accela ChemBio Inc.

2,2,2-Trifluoro-N-(4-isopropylphenyl)acetamide

  • MDL number MFCD00591832
  • Cas Number : 75999-61-0
  • Molecular Weight : 231.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018388&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018388.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart