AC5372911903 Accela ChemBio Inc.

N-(2-Benzoyl-4-chlorophenyl)-3,4-difluorobenzamide

  • MDL number MFCD02084064
  • Cas Number : 312923-12-9
  • Molecular Weight : 371.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018389&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018389.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart