AC5372911904 Accela ChemBio Inc.

N-(4-Acetamidophenyl)-2-ethoxyacetamide

  • MDL number MFCD03929376
  • Cas Number : 600140-39-4
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018390&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018390.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart