AC5372911905 Accela ChemBio Inc.

N-(4-Cyanophenyl)-3,4-difluorobenzamide

  • MDL number MFCD09951416
  • Cas Number : 1007802-68-7
  • Molecular Weight : 258.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018391&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018391.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart