AC5372911907 Accela ChemBio Inc.

N-[1-(Hydroxymethyl)propyl]-4-methylbenzenesulfonamide

  • MDL number MFCD00458993
  • Cas Number : 304460-78-4
  • Molecular Weight : 243.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018393&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018393.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart