AC5372911914 Accela ChemBio Inc.

2-Bromo-4-(bromomethyl)benzonitrile

  • MDL number MFCD09261043
  • Cas Number : 89892-38-6
  • Molecular Weight : 274.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018414&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018414.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart