AC5372911922 Accela ChemBio Inc.

2-Aminobenzothiazole-6-carboxylic Acid Hydrochloride

  • MDL number MFCD01333078
  • Cas Number : 18330-76-2
  • Molecular Weight : 230.67138

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018451&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018451.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart