AC5372911937 Accela ChemBio Inc.

3-(Aminomethyl)-5-propylpyrazole

  • MDL number MFCD08700610
  • Cas Number : 1093415-70-3
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018552&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018552.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart