AC5372911938 Accela ChemBio Inc.

3-(Aminomethyl)-5-(4-pyridyl)pyrazole

  • MDL number MFCD22205846
  • Cas Number : 1004303-48-3
  • Molecular Weight : 174.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018553&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018553.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart