AC5372911964 Accela ChemBio Inc.

2-Cyano-3-phenylpropionic Acid

  • MDL number MFCD00205073
  • Cas Number : 5331-42-0
  • Molecular Weight : 175.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018695&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018695.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart