AC5372911965 Accela ChemBio Inc.

4-Fluoro-2-[(methylamino)methyl]phenol

  • MDL number MFCD21641325
  • Cas Number : 1363166-05-5
  • Molecular Weight : 155.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018704&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018704.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart