AC5372911966 Accela ChemBio Inc.

2-[(Ethylamino)methyl]-4-fluorophenol

  • MDL number MFCD21641326
  • Cas Number : 1363166-27-1
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018705&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY018705.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart