AC5372911967 Accela ChemBio Inc.

Spiro[1,2,3,4-tetrahydroquinazoline-2,1'-cyclobutane]-4-one

  • MDL number MFCD21641327
  • Cas Number : 1363165-92-7
  • Molecular Weight : 188.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018717&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018717.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart