AC5372911974 Accela ChemBio Inc.

2,4-Dichloro-N-[4-(2,4-dichlorobenzoyloxy)-2-methylphenyl]benzamide

  • MDL number MFCD21641332
  • Cas Number : 1363166-08-8
  • Molecular Weight : 469.14482

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018775&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018775.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart