AC5372911977 Accela ChemBio Inc.

Isopropyl 4-Hydroxy-3,5-dimethoxybenzoate

  • MDL number MFCD21641333
  • Cas Number : 858131-80-3
  • Molecular Weight : 240.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name isopropyl 4-hydroxy-3,5-dimethoxybenzoate
Inchi Code 1S/C12H16O5/c1-7(2)17-12(14)8-5-9(15-3)11(13)10(6-8)16-4/h5-7,13H,1-4H3
Inchi Key UFDMGVMGWSSNPP-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to Brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018779&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY018779.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart