AC5372911991 Accela ChemBio Inc.

N-(3-Chloro-4-methylphenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD01185222
  • Cas Number : 64694-83-3
  • Molecular Weight : 237.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018841&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018841.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart