AC5372911992 Accela ChemBio Inc.

5-Bromo-2-naphthyl 2-(6-Methoxy-2-naphthyl)propanoate

  • MDL number MFCD21641336
  • Cas Number : 1363166-31-7
  • Molecular Weight : 435.30986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018842&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018842.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart