AC5372911993 Accela ChemBio Inc.

N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-4-pyrazolyl)butane-1-sulfonamide

  • MDL number MFCD01594036
  • Cas Number : 895767-41-6
  • Molecular Weight : 323.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018845&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018845.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart