AC5372912007 Accela ChemBio Inc.

2-Methoxy-5-sulfamoylbenzoic Acid

  • MDL number MFCD00129997
  • Cas Number : 22117-85-7
  • Molecular Weight : 231.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018905&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018905.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart