AC5372912028 Accela ChemBio Inc.

1H-Benzo[d]azepin-2(3H)-one

  • MDL number MFCD01101239
  • Cas Number : 19301-09-8
  • Molecular Weight : 159.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018989&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018989.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart