AC5372912040 Accela ChemBio Inc.

O-[(Ethoxycarbonyl)cyanomethylenamino]-N,N,N',N'-tetramethyluronium Tetrafluoroborate

  • MDL number MFCD00192127
  • Cas Number : 136849-72-4
  • Molecular Weight : 328.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019065&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY019065.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart