AC5372912046 Accela ChemBio Inc.

2-Chloro-3-methyl-4-pyridinecarboxylic Acid

  • MDL number MFCD16610679
  • Cas Number : 133928-73-1
  • Molecular Weight : 171.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019118&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019118.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart