AC5372912050 Accela ChemBio Inc.

Methyl 2-Amino-4-morpholinobenzoate

  • MDL number MFCD22192343
  • Cas Number : 404010-84-0
  • Molecular Weight : 236.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019138&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019138.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart