AC5372912051 Accela ChemBio Inc.

5-Acetyl-4-Chloro-7H-pyrrolo[2,3-d]pyrimidine

  • MDL number MFCD22205848
  • Cas Number : 1363381-59-2
  • Molecular Weight : 195.60574

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019146&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019146.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart