AC5372912052 Accela ChemBio Inc.

N-Benzyl-3-bromoaniline

  • MDL number MFCD11145017
  • Cas Number : 213814-61-0
  • Molecular Weight : 262.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019152&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019152.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart