AC5372912053 Accela ChemBio Inc.

alpha-(1-Naphthyl)benzylamine

  • MDL number MFCD00168077
  • Cas Number : 2936-63-2
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019153&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019153.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart