AC5372912070 Accela ChemBio Inc.

N-[3-(1-Imidazolyl)propyl]-2,4-dichlorobenzamide

  • MDL number MFCD01920187
  • Cas Number : 349092-52-0
  • Molecular Weight : 298.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-dichloro-N-[3-(1H-imidazol-1-yl)propyl]benzamide
Inchi Code 1S/C13H13Cl2N3O/c14-10-2-3-11(12(15)8-10)13(19)17-4-1-6-18-7-5-16-9-18/h2-3,5,7-9H,1,4,6H2,(H,17,19)
Inchi Key GUTLIVUMYAQCOK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019219&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019219.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart