AC5372912074 Accela ChemBio Inc.

N-(2,4-Dichlorophenyl)ethanesulfonamide

  • MDL number MFCD03383712
  • Cas Number : 56682-60-1
  • Molecular Weight : 254.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019226&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019226.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart