AC5372912076 Accela ChemBio Inc.

N-(2-Benzoyl-4-chlorophenyl)butane-1-sulfonamide

  • MDL number MFCD02317335
  • Cas Number : 392305-35-0
  • Molecular Weight : 351.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(2-benzoyl-4-chlorophenyl)-1-butanesulfonamide
Inchi Code 1S/C17H18ClNO3S/c1-2-3-11-23(21,22)19-16-10-9-14(18)12-15(16)17(20)13-7-5-4-6-8-13/h4-10,12,19H,2-3,11H2,1H3
Inchi Key YIGQVHGBAPUGIY-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019228&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019228.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart