AC5372912078 Accela ChemBio Inc.

3-(Boc-amino)-3-(4-methoxyphenyl)-1-propanol

  • MDL number MFCD18907511
  • Cas Number : 96363-26-7
  • Molecular Weight : 281.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 3-hydroxy-1-(4-methoxyphenyl)propylcarbamate
Inchi Code 1S/C15H23NO4/c1-15(2,3)20-14(18)16-13(9-10-17)11-5-7-12(19-4)8-6-11/h5-8,13,17H,9-10H2,1-4H3,(H,16,18)
Inchi Key KIILCIAJJCYALF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019233&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019233.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart