AC5372912094 Accela ChemBio Inc.

Methyl N-Boc-4-(trifluoroacetylamino)piperidine-4-carboxylate

  • MDL number MFCD22192346
  • Cas Number : 1375064-57-5
  • Molecular Weight : 354.3221496

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019285&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019285.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart