AC5372912095 Accela ChemBio Inc.

alpha-(4-Biphenylyl)benzylamine

  • MDL number MFCD11848907
  • Cas Number : 91487-88-6
  • Molecular Weight : 259.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019288&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019288.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart