AC5372912106 Accela ChemBio Inc.

2,4-Dichloro-6-(methylthio)-1,3,5-triazine

  • MDL number MFCD01679001
  • Cas Number : 13705-05-0
  • Molecular Weight : 196.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form White to yellow solid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019326&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019326.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart