AC5372912107 Accela ChemBio Inc.

3,4-Difluoro-N-(2-hydroxypropyl)benzamide

  • MDL number MFCD11936927
  • Cas Number : 1156264-10-6
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019335&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019335.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart