AC5372912113 Accela ChemBio Inc.

2-Fluoro-N-(6-methyl-2-pyridyl)benzenesulfonamide

  • MDL number MFCD12635817
  • Cas Number : 1136813-84-7
  • Molecular Weight : 266.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-N-(6-methyl-2-pyridinyl)benzenesulfonamide
Inchi Code 1S/C12H11FN2O2S/c1-9-5-4-8-12(14-9)15-18(16,17)11-7-3-2-6-10(11)13/h2-8H,1H3,(H,14,15)
Inchi Key QPRYMOBGMZPONZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. RT
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019357&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019357.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart