AC5372912118 Accela ChemBio Inc.

3-Amino-4-methyl-N-(4-methyl-2-pyridyl)benzamide

  • MDL number MFCD11131813
  • Cas Number : 1019398-93-6
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019377&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019377.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart