AC5372912120 Accela ChemBio Inc.

2-(Methylthio)-5,6,7,8-tetrahydropyrido[3,2-d]pyrimidine Hydrochloride

  • MDL number MFCD22192347
  • Cas Number : 1375064-63-3
  • Molecular Weight : 217.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(methylsulfanyl)-5,6,7,8-tetrahydropyrido[3,2-d]pyrimidine hydrochloride
Inchi Code 1S/C8H11N3S.ClH/c1-12-8-10-5-7-6(11-8)3-2-4-9-7;/h5,9H,2-4H2,1H3;1H
Inchi Key MPUMFJCLDZVTGY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019384&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY019384.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart