AC5372912122 Accela ChemBio Inc.

Ethyl 2-Amino-4-(1-pyrazolyl)benzoate

  • MDL number MFCD22205850
  • Cas Number : 1375064-68-8
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019387&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019387.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart