AC5372912130 Accela ChemBio Inc.

alpha-(4-Hydroxyphenyl)benzylamine

  • MDL number MFCD06655632
  • Cas Number : 81123-45-7
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY019424&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY019424.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart